Learn More
Biodesign Inc of Ny Sieve Holder, two piece plastic with metal tightener, 3 inches (7.6cm) diameter
Catalog No. NC2684791
Encompass
Sieve Holder used to tightly hold CellMicroSieve filters to form a funnel through which extraction liquid can easily be collected
Catalog No. NC2684791 Supplier Biodesign Inc of Ny Supplier No. F3
Sieve Holder used to tightly hold CellMicroSieve filters to form a funnel through which extraction liquid can easily be collected