Learn More
Fritsch Milling And Sizing Inc Sieve Insert P-15 0.5mm Trapez Perf., Chromium-Free Steel
SDP
Catalog No. 501534462
Encompass
Sieve insert P-15 0.5mm trapez perf., chromium-free steel
Sieve inserts determine the fineness of a sample.
Sieve inserts with trapezoidal perforation and square perforations are offered. Sieve inserts with trapezoidal perforations improve size reduction through extra shearing action. Sieve inserts with square perforation are recommended for the grinding of brittle materials and materials of medium fineness with a narrow grain band.
The sieve inserts made of chromium free steel ST12 are especially recommended for the sample preparation according to RoHS and WEEE (for the poof of hexavalent chromium) and for the XRF- analysis.
For all other applications sieve inserts made of stainless steel may be used since these are harder and more resilient against abrasion.
Catalog No. 50-153-4462 Supplier Fritsch Milling And Sizing Inc Supplier No. 45.1211.09
Sieve insert P-15 0.5mm trapez perf., chromium-free steel
Sieve inserts determine the fineness of a sample.
Sieve inserts with trapezoidal perforation and square perforations are offered. Sieve inserts with trapezoidal perforations improve size reduction through extra shearing action. Sieve inserts with square perforation are recommended for the grinding of brittle materials and materials of medium fineness with a narrow grain band.
The sieve inserts made of chromium free steel ST12 are especially recommended for the sample preparation according to RoHS and WEEE (for the poof of hexavalent chromium) and for the XRF- analysis.
For all other applications sieve inserts made of stainless steel may be used since these are harder and more resilient against abrasion.