Learn More
Fritsch Milling And Sizing Inc Sieve Pan with Outlet 100/40mm Stainless Steel
SDP
Catalog No. 501534173
Encompass
For wet sieving with test sieves 100 mm dia.
The sieve pan made of stainless steel with outlet completes the sieve stack.
Catalog No. 50-153-4173 Supplier Fritsch Milling And Sizing Inc Supplier No. 31.1140.00
For wet sieving with test sieves 100 mm dia.
The sieve pan made of stainless steel with outlet completes the sieve stack.