Learn More
Fritsch Milling And Sizing Inc Sieve Ring P-14 Premium Line 2 mm Round Perf.
SDP
Catalog No. 501533961
Encompass
Sieve ring 2 mm trapezoidal perforation with reinforced edges made of stainless steel
Sieve rings determine the final fineness of the sample.
Sieve rings with reinforced edges and trapezoidal perforation are perfect for fast comminution of medium-hard to soft materials, the trapezoidal perforation provides additional shearing effects.
Additionally are sieve rings with reinforced edges with round perforation available which are perfect for brittle materials and medium fineness with narrow particle size range.
Sieve rings are also available in further perforations.
Catalog No. 50-153-3961 Supplier Fritsch Milling And Sizing Inc Supplier No. 14.4361.00
Sieve ring 2 mm trapezoidal perforation with reinforced edges made of stainless steel
Sieve rings determine the final fineness of the sample.
Sieve rings with reinforced edges and trapezoidal perforation are perfect for fast comminution of medium-hard to soft materials, the trapezoidal perforation provides additional shearing effects.
Additionally are sieve rings with reinforced edges with round perforation available which are perfect for brittle materials and medium fineness with narrow particle size range.
Sieve rings are also available in further perforations.