Learn More
Fritsch Milling And Sizing Inc Sieve Ring P-14 Cl Stainl. Steel 0.5 mm Trapezoidal Perf.
SDP
Catalog No. 501534419
Encompass
Sieve ring P-14 cl stainl. steel 0.5 mm trapezoidal perf.
Sieve rings determine the end fineness of the sample.
Sieve rings with trapezoidal perforations and sieve rings with round perforations are available.
Sieve rings with trapezoidal perforations improve size reduction through extra shearing action.
Sieve rings with round perforation are recommended for reduction of brittle and medium fine materials with a smaller grain band.
Sieve rings with reinforced edges for heavy duty size reduction are also offered.
Catalog No. 50-153-4419 Supplier Fritsch Milling And Sizing Inc Supplier No. 44.1050.10
Sieve ring P-14 cl stainl. steel 0.5 mm trapezoidal perf.
Sieve rings determine the end fineness of the sample.
Sieve rings with trapezoidal perforations and sieve rings with round perforations are available.
Sieve rings with trapezoidal perforations improve size reduction through extra shearing action.
Sieve rings with round perforation are recommended for reduction of brittle and medium fine materials with a smaller grain band.
Sieve rings with reinforced edges for heavy duty size reduction are also offered.