Learn More
Fritsch Milling And Sizing Inc Sieve Ring P-14 Cl Stainl. Steel 1 mm Trapezoidal Perf.
SDP
Catalog No. 501534422
Encompass
Sieve ring P-14 cl stainl. steel 1 mm trapezoidal perf.
Sieve rings determine the end fineness of the sample.
Sieve rings with trapezoidal perforations and sieve rings with round perforations are offered.
Sieve rings with trapezoidal perforations improve size reduction through extra shearing action.
Sieve rings with round perforations are recommended for grinding brittle and materials of medium fineness with small grain bands.
Also sieve rings with reinforced edges for heavy duty grinding are offered.
Catalog No. 50-153-4422 Supplier Fritsch Milling And Sizing Inc Supplier No. 44.1100.10
Sieve ring P-14 cl stainl. steel 1 mm trapezoidal perf.
Sieve rings determine the end fineness of the sample.
Sieve rings with trapezoidal perforations and sieve rings with round perforations are offered.
Sieve rings with trapezoidal perforations improve size reduction through extra shearing action.
Sieve rings with round perforations are recommended for grinding brittle and materials of medium fineness with small grain bands.
Also sieve rings with reinforced edges for heavy duty grinding are offered.