Learn More
Fritsch Milling And Sizing Inc Sieve Ring P-14 Cl Stainl. Steel 4 mm Round Perforation
SDP
Catalog No. 501534440
Encompass
Sieve ring P-14 cl stainl. steel 4 mm round perforation
Sieve rings determine the end fineness of the sample.
Sieve rings with round and trapezoidal perforations are offered.
Sieve rings with round perforations are recommended for grinding brittle and materials of medium fineness with small grain bands.
Sieve rings with trapezoidal perforations improve size reduction through extra shearing action.
Catalog No. 50-153-4440 Supplier Fritsch Milling And Sizing Inc Supplier No. 44.2400.10
Sieve ring P-14 cl stainl. steel 4 mm round perforation
Sieve rings determine the end fineness of the sample.
Sieve rings with round and trapezoidal perforations are offered.
Sieve rings with round perforations are recommended for grinding brittle and materials of medium fineness with small grain bands.
Sieve rings with trapezoidal perforations improve size reduction through extra shearing action.