Learn More
Fritsch Milling And Sizing Inc Sieve Ring P-14 Cl Stl. Steel 0.08 mm Trapezoidal Perf.
SDP
Catalog No. 501534413
Encompass
Sieve ring P-14 cl stl. steel 0.08 mm trapezoidal perf.
Sieve rings determine the final fineness of the sample.
Sieve rings with round and trapezoidal perforations are available.
Sieve rings with trapezoidal perforations improve size reduction through extra shearing action.
Sieve rings with round perforation are recommended for reduction of brittle materials and medium fine materials with a smaller grain band.
Sieve rings for reinforced edges for heavy duty size reduction are also offered.
Catalog No. 50-153-4413 Supplier Fritsch Milling And Sizing Inc Supplier No. 44.1000.10
Sieve ring P-14 cl stl. steel 0.08 mm trapezoidal perf.
Sieve rings determine the final fineness of the sample.
Sieve rings with round and trapezoidal perforations are available.
Sieve rings with trapezoidal perforations improve size reduction through extra shearing action.
Sieve rings with round perforation are recommended for reduction of brittle materials and medium fine materials with a smaller grain band.
Sieve rings for reinforced edges for heavy duty size reduction are also offered.