Learn More
Fritsch Milling And Sizing Inc Sieve Ring Pure Titanium 0.08 mm Trapezoidal Perf., P-14 Pl
SDP
Catalog No. 501533968
Encompass
Sieve ring 0.08 mm trapezoidal perforation for PULVERISETTE 14 premium line with reinforced edges, pure titanium for heavy-metal and iron-free grinding and sample preparation according to RoHS.
Sieve rings determine the final fineness of the sample.
Sieve rings with reinforced edges and trapezoidal perforation are perfect for fast comminution of medium-hard to soft materials, the trapezoidal perforation provides additional shearing effects. 
Catalog No. 50-153-3968 Supplier Fritsch Milling And Sizing Inc Supplier No. 14.4441.32
Sieve ring 0.08 mm trapezoidal perforation for PULVERISETTE 14 premium line with reinforced edges, pure titanium for heavy-metal and iron-free grinding and sample preparation according to RoHS.
Sieve rings determine the final fineness of the sample.
Sieve rings with reinforced edges and trapezoidal perforation are perfect for fast comminution of medium-hard to soft materials, the trapezoidal perforation provides additional shearing effects. 