Learn More
Grainger Sieve Shaker Conversion Kit
Catalog No. 502768067
Encompass_Preferred
The Grainger website offers electric sieve shakers for laboratory use, designed to separate materials by particle size with standard test sieves. They feature variable speed controls, built-in timers, and durable metal construction. Models include the W.S. Tyler Ro-Tap with 8-inch and 12-inch capacities and the Gilson SS-15 with a 15-inch capacity. These shakers enable efficient, reproducible sieving for soil testing, powder classification, and quality control, with customizable settings for different materials and sizes.
Catalog No. 50-276-8067 Supplier Grainger Supplier No. 16Y906
Add to Cart
Add to Cart
The Grainger website offers electric sieve shakers for laboratory use, designed to separate materials by particle size with standard test sieves. They feature variable speed controls, built-in timers, and durable metal construction. Models include the W.S. Tyler Ro-Tap with 8-inch and 12-inch capacities and the Gilson SS-15 with a 15-inch capacity. These shakers enable efficient, reproducible sieving for soil testing, powder classification, and quality control, with customizable settings for different materials and sizes.