Learn More
Gilson Company Inc 200mm diameter Air Jet Test Sieve, No. 200
Catalog No. NC9002947
Encompass_Preferred
200mm diameter Air Jet Test Sieve has stainless steel woven-wire cloth with No. 200 (75µm) opening size and stainless-steel frame and is fitted with special sealing gasket. Features: Specially designed for Micron Air Jet Sieves. Compliant with ASTM E11 requirements. Included Items: Verification Services provide NIST traceable documentation of opening sizes.
Catalog No. NC9002947 Supplier Gilson Company Inc Supplier No. AJA218
200mm diameter Air Jet Test Sieve has stainless steel woven-wire cloth with No. 200 (75µm) opening size and stainless-steel frame and is fitted with special sealing gasket. Features: Specially designed for Micron Air Jet Sieves. Compliant with ASTM E11 requirements. Included Items: Verification Services provide NIST traceable documentation of opening sizes.