Learn More
Gilson Company Inc 200mm diameter Air Jet Test Sieve, No. 325
Catalog No. NC9002948
Encompass_Preferred
200mm diameter Air Jet Test Sieve has stainless steel woven-wire cloth with No. 325 (45µm) opening size and stainless-steel frame and is fitted with special sealing gasket. Features: Specially designed for Micron Air Jet Sieves. Compliant with ASTM E11 requirements. Included Items: Verification Services provide NIST traceable documentation of opening sizes.
Catalog No. NC9002948 Supplier Gilson Company Inc Supplier No. AJA214
200mm diameter Air Jet Test Sieve has stainless steel woven-wire cloth with No. 325 (45µm) opening size and stainless-steel frame and is fitted with special sealing gasket. Features: Specially designed for Micron Air Jet Sieves. Compliant with ASTM E11 requirements. Included Items: Verification Services provide NIST traceable documentation of opening sizes.