Learn More
Gilson Company Inc No. 200 Mesh Sieve, All Stainless, Full Height, 8" Diameter
Catalog No. NC0306898
Encompass_Preferred
8in (203mm) diameter ASTM E11 Test Sieve is constructed with a No. 200 (75µm) stainless steel woven-wire cloth opening size, and a full-height stainless steel frame. The overall height is 2-5/8in and the stacked height is 2-1/8in. This sieve is optionally available fitted with #35 (500µm) stainless steel backing cloth for enhanced durability.
Catalog No. NC0306898 Supplier Gilson Company Inc Supplier No. V8SF #200
8in (203mm) diameter ASTM E11 Test Sieve is constructed with a No. 200 (75µm) stainless steel woven-wire cloth opening size, and a full-height stainless steel frame. The overall height is 2-5/8in and the stacked height is 2-1/8in. This sieve is optionally available fitted with #35 (500µm) stainless steel backing cloth for enhanced durability.