Learn More
Gilson Company Inc 1L Sieving Aid
Catalog No. NC0933791
Encompass_Preferred
Sieving Aid is a very fine (5—7 um), high-purity silicon dioxide powder. When mixed with fine samples (2% by weight is typical), Sieving Aid keeps material free-flowing and prevents agglomeration from static attraction or high humidity. It also acts as a dehydrating agent for samples.
Catalog No. NC0933791 Supplier Gilson Company Inc Supplier No. SSA58
Sieving Aid is a very fine (5—7 um), high-purity silicon dioxide powder. When mixed with fine samples (2% by weight is typical), Sieving Aid keeps material free-flowing and prevents agglomeration from static attraction or high humidity. It also acts as a dehydrating agent for samples.