Learn More
Medchemexpress LLC Sifuvirtide acetate | 4727.98 (free base) | 10 MG
SDP
Catalog No. 5000423788
Encompass_Preferred
Sifuvirtide (SFT) acetate is a powerful HIV fusion inhibitor that prevents HIV-1 mediated cell fusion in a dose-dependent manner. It demonstrates high potency against both primary and laboratory-adapted HIV-1 isolates across various genotypes, making it suitable for anti-HIV drug research.
  • Potent HIV fusion inhibitor.
  • Inhibits HIV-1 mediated cell fusion in a dose-dependent manner.
  • Highly potent against infection by primary and laboratory-adapted HIV-1 isolates of multiple genotypes.
  • Can be used in the research of anti-HIV drugs.
Catalog No. 50-004-23788 Supplier Medchemexpress LLC Supplier No. HYP10517A10MG
Sifuvirtide (SFT) acetate is a powerful HIV fusion inhibitor that prevents HIV-1 mediated cell fusion in a dose-dependent manner. It demonstrates high potency against both primary and laboratory-adapted HIV-1 isolates across various genotypes, making it suitable for anti-HIV drug research.
  • Potent HIV fusion inhibitor.
  • Inhibits HIV-1 mediated cell fusion in a dose-dependent manner.
  • Highly potent against infection by primary and laboratory-adapted HIV-1 isolates of multiple genotypes.
  • Can be used in the research of anti-HIV drugs.