Learn More
Medchemexpress LLC Sifuvirtide acetate | 857094-21-4 | 99.8% | 4727.98 | 5MG
SDP
Catalog No. 5000423726
Encompass_Preferred
Sifuvirtide acetate is a potent HIV fusion inhibitor. It inhibits HIV-1 mediated cell fusion in a dose-dependent manner and is highly potent against infection by primary and laboratory-adapted HIV-1 isolates of multiple genotypes. This product is suitable for anti-HIV drug research.
  • Potent HIV fusion inhibitor
  • Inhibits HIV-1 mediated cell fusion in a dose-dependent manner
  • Highly potent against infection by primary and laboratory-adapted HIV-1 isolates of multiple genotypes
  • Suitable for anti-HIV drug research
  • Sealed storage, away from moisture
  • Powder storage: -80°C for 2 years; -20°C for 1 year
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
Catalog No. 50-004-23726 Supplier Medchemexpress LLC Supplier No. HYP10517A5MG
Sifuvirtide acetate is a potent HIV fusion inhibitor. It inhibits HIV-1 mediated cell fusion in a dose-dependent manner and is highly potent against infection by primary and laboratory-adapted HIV-1 isolates of multiple genotypes. This product is suitable for anti-HIV drug research.
  • Potent HIV fusion inhibitor
  • Inhibits HIV-1 mediated cell fusion in a dose-dependent manner
  • Highly potent against infection by primary and laboratory-adapted HIV-1 isolates of multiple genotypes
  • Suitable for anti-HIV drug research
  • Sealed storage, away from moisture
  • Powder storage: -80°C for 2 years; -20°C for 1 year
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)