Learn More
Medchemexpress LLC Sifuvirtide acetate | 99.8% | 4727.98 | 25 MG
SDP
Catalog No. 5000426800
Encompass_Preferred
Sifuvirtide acetate is a potent HIV fusion inhibitor. It inhibits HIV-1 mediated cell fusion in a dose-dependent manner and is highly potent against infection by primary and laboratory-adapted HIV-1 isolates of multiple genotypes. This product can be used in the research of anti-HIV drugs.
  • Potent HIV fusion inhibitor
  • Inhibits HIV-1 mediated cell fusion in a dose-dependent manner
  • Highly potent against infection by primary and laboratory-adapted HIV-1 isolates of multiple genotypes
  • Used for anti-HIV drug research
Catalog No. 50-004-26800 Supplier Medchemexpress LLC Supplier No. 25MGHYP10517A
Sifuvirtide acetate is a potent HIV fusion inhibitor. It inhibits HIV-1 mediated cell fusion in a dose-dependent manner and is highly potent against infection by primary and laboratory-adapted HIV-1 isolates of multiple genotypes. This product can be used in the research of anti-HIV drugs.
  • Potent HIV fusion inhibitor
  • Inhibits HIV-1 mediated cell fusion in a dose-dependent manner
  • Highly potent against infection by primary and laboratory-adapted HIV-1 isolates of multiple genotypes
  • Used for anti-HIV drug research