Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED siGENOME Non-Targeting siRNA Control Pool #1 | 20 nmol
Catalog No. NC1579866
Encompass
siGENOME Non-Targeting siRNA Control Pools are highly functional, chemically synthesized, and mostly unmodified negative control siRNA duplexes for experiments in human mouse and rat cells. siGENOME Non-Targeting Control Pools are each a pool of four siRNAs, allowing the effective concentration of each siRNA to be reduced, and thus further minimize off-target effects. siGENOME Non-Targeting Control siRNAs were designed to have a minimum of 4 mismatches to all human, mouse and rat genes, and confirmed to have minimal targeting by genomewide microarray analysis. siGENOME Non-Targeting Pool #1 is comprised of Non-Targeting siRNAs #1 through 4.
Catalog No. NC1579866 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. D0012061320
siGENOME Non-Targeting siRNA Control Pools are highly functional, chemically synthesized, and mostly unmodified negative control siRNA duplexes for experiments in human mouse and rat cells. siGENOME Non-Targeting Control Pools are each a pool of four siRNAs, allowing the effective concentration of each siRNA to be reduced, and thus further minimize off-target effects. siGENOME Non-Targeting Control siRNAs were designed to have a minimum of 4 mismatches to all human, mouse and rat genes, and confirmed to have minimal targeting by genomewide microarray analysis. siGENOME Non-Targeting Pool #1 is comprised of Non-Targeting siRNAs #1 through 4.