Learn More
Aviva Systems Biology Corp SIGIRR Antibody - C-terminal region (ARP49664_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49664_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIGIRR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SIGIRR. Alternate names: MGC110992, TIR8ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-60142