Learn More
Novus Biologicals™ SIGIRR Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 59307 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56890 |
| Gene Alias | single Ig IL-1-related receptor, Single Ig IL-1 R-related molecule, single immunoglobulin and toll-interleukin 1 receptor (TIR) domain, single immunoglobulin domain IL1R1 related, Single immunoglobulin domain-containing IL1R-related protein, TIR8MGC110992, Toll/interleukin-1 receptor 8 |
| Gene Symbol | SIGIRR |
| Quantity | 0.1 mg |
| Research Category | Innate Immunity |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.