Learn More
AdipoGen SIGIRR hFc h rec
SDP
Supplier:  AdipoGen AG40A0093TC050
Encompass_Preferred
Protein. Signal peptide and extracellular domain of human SIGIRR aa 1-118 are fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.06EU/µg purified protein LAL test Lonza. Lyophilized from 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. SIGIRR acts as a negative regulator of the Toll-like and IL-1R receptor signaling pathways. Attenuates the recruitment of receptor-proximal signaling components to the TLR4 receptor, probably through a TIR-TIR domain interaction with TLR4.
Catalog No. 50-256-3003