Learn More
ACROBiosystems ACROBiosystems Mouse Siglec-10 Protein, His Tag
Supplier:  ACROBiosystems SI0M52H7100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse Siglec-10, His Tag (SI0-M52H7) is expressed from human 293 cells (HEK293). It contains AA Met 18 - Lys 543 (Accession # Q80ZE3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.6 kDa.
Catalog No. 50-195-6548