Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Siglec-10 Protein, His,Avitag™
Supplier:  ACROBiosystems SI0H82E3200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human Siglec-10, His,Avitag (SI0-H82E3) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Accession # Q96LC7-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 61.8 kDa. The protein migrates as 65-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Siglec-10, His,Avitag (Cat. No. SI0-H82E3) at 5 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Anti-Human Siglec-10 MAb, human IgG with a linear range of 0.6-20 ng/mL (QC tested).
Catalog No. 50-195-6543