Learn More
Medchemexpress LLC Siglec-10 Human Bi 100ug
SDP
Catalog No. 5000181937
Encompass_Preferred
Siglec-10 protein is thought to be a putative adhesion molecule that mediates sialic acid-dependent cell binding preferentially selecting -2 3- or -2 6-linked sialic acid Its sialic acid recognition site may be masked by cis interactions Siglec-10 Protein Human (Biotinylated HEK293 Fc-Avi) is the recombinant human-derived Siglec-10 protein expressed by HEK293 with C-Avi C-hFc labeled tag
Catalog No. 50-001-81937 Supplier Medchemexpress LLC Supplier No. HYP72411100UG
Add to Cart
Add to Cart
Siglec-10 protein is thought to be a putative adhesion molecule that mediates sialic acid-dependent cell binding preferentially selecting -2 3- or -2 6-linked sialic acid Its sialic acid recognition site may be masked by cis interactions Siglec-10 Protein Human (Biotinylated HEK293 Fc-Avi) is the recombinant human-derived Siglec-10 protein expressed by HEK293 with C-Avi C-hFc labeled tag