Learn More
Medchemexpress LLC Siglec-10 Human Bi 20ug
SDP
Catalog No. 5000181938
Encompass_Preferred
Siglec-10 protein is thought to be a putative adhesion molecule that mediates sialic acid-dependent cell binding preferentially selecting -2 3- or -2 6-linked sialic acid Its sialic acid recognition site may be masked by cis interactions Siglec-10 Protein Human (Biotinylated HEK293 Fc-Avi) is the recombinant human-derived Siglec-10 protein expressed by HEK293 with C-Avi C-hFc labeled tag
Catalog No. 50-001-81938 Supplier Medchemexpress LLC Supplier No. HYP7241120UG
Siglec-10 protein is thought to be a putative adhesion molecule that mediates sialic acid-dependent cell binding preferentially selecting -2 3- or -2 6-linked sialic acid Its sialic acid recognition site may be masked by cis interactions Siglec-10 Protein Human (Biotinylated HEK293 Fc-Avi) is the recombinant human-derived Siglec-10 protein expressed by HEK293 with C-Avi C-hFc labeled tag