Learn More
Genscript Corporation Siglec-15/CD33L3 hFc Chimera
Catalog No. 5000058273
Encompass
Siglec-15 is a transmembrane glycoprotein in the Siglec family of sialic acid-binding immune regulatory molecules Mature human Siglec-15 consists of a 244 amino acid extracellular domain (ECD) with two Ig-like domains a 21 aa transmembrane segment and a 44 aa cytoplasmic domain Siglec-15 is a potential therapeutic target for osteoporosis and plays a conserved regulatory role in the immune system of vertebrates
Catalog No. 50-000-58273 Supplier Genscript Corporation Supplier No. Z05820500
Siglec-15 is a transmembrane glycoprotein in the Siglec family of sialic acid-binding immune regulatory molecules Mature human Siglec-15 consists of a 244 amino acid extracellular domain (ECD) with two Ig-like domains a 21 aa transmembrane segment and a 44 aa cytoplasmic domain Siglec-15 is a potential therapeutic target for osteoporosis and plays a conserved regulatory role in the immune system of vertebrates