Learn More
ACROBiosystems ACROBiosystems Cynomolgus Siglec-15 / CD33L3 Protein, Fc Tag
Catalog No. 501956790
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus Siglec-15, Fc Tag (SG5-C5253) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession # A0A2K5UY47-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.7 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus Siglec-15, Fc Tag (Cat. No. SG5-C5253) at 5 μg/mL (100 μL/well) can bind Mouse Human chimeric MAb (5G12, Human IgG1) with a linear range of 0.4-6 ng/mL (QC tested).;Immobilized Cynomolgus Siglec-15, Fc Tag (Cat. No. SG5-C5253) at 10 μg/mL (100 μL/well) on Diamond Protein A Protein, His Tag precoated (0.5 μg/well) plate, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 0.078-1.25 μg/mL (Routinely tested).
Catalog No. 50-195-6790 Supplier ACROBiosystems Supplier No. SG5C52531MG
Recombinant Protein;1MG;Cynomolgus Siglec-15, Fc Tag (SG5-C5253) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession # A0A2K5UY47-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.7 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus Siglec-15, Fc Tag (Cat. No. SG5-C5253) at 5 μg/mL (100 μL/well) can bind Mouse Human chimeric MAb (5G12, Human IgG1) with a linear range of 0.4-6 ng/mL (QC tested).;Immobilized Cynomolgus Siglec-15, Fc Tag (Cat. No. SG5-C5253) at 10 μg/mL (100 μL/well) on Diamond Protein A Protein, His Tag precoated (0.5 μg/well) plate, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 0.078-1.25 μg/mL (Routinely tested).