Learn More
ACROBiosystems ACROBiosystems Cynomolgus Siglec-15 / CD33L3 Protein, His Tag
Supplier:  ACROBiosystems SG5C52H6100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus Siglec-15, His Tag (SG5-C52H6) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession # A0A2K5UY47-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.1 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus Siglec-15, His Tag (Cat. No. SG5-C52H6) at 5 μg/mL (100 μL/well) can bind Mouse Human chimeric MAb (5G12, Human IgG1) with a linear range of 0.3-2 ng/mL (QC tested).
Catalog No. 50-195-6899