Learn More
ACROBiosystems ACROBiosystems Human Siglec-15 / CD33L3 Protein, His Tag
Catalog No. 501957032
Encompass_Preferred
Recombinant Protein;100UG;Human Siglec-15, His Tag (SG5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession # Q6ZMC9-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.4 kDa. The protein migrates as 32-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-15, His Tag (Cat. No. SG5-H52H3) at 5 μg/mL (100 μL/well) can bind Mouse Human chimeric MAb (5G12, Human IgG1) with a linear range of 0.3-2 ng/mL (QC tested).;Immobilized Human Siglec-15, His Tag (Cat. No. SG5-H52H3) at 10 μg/mL (100 μL/well) on an Nickel Coated plate, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 0.078-5 μg/mL (Routinely tested).
Catalog No. 50-195-7032 Supplier ACROBiosystems Supplier No. SG5H52H3100UG
Recombinant Protein;100UG;Human Siglec-15, His Tag (SG5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession # Q6ZMC9-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.4 kDa. The protein migrates as 32-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-15, His Tag (Cat. No. SG5-H52H3) at 5 μg/mL (100 μL/well) can bind Mouse Human chimeric MAb (5G12, Human IgG1) with a linear range of 0.3-2 ng/mL (QC tested).;Immobilized Human Siglec-15, His Tag (Cat. No. SG5-H52H3) at 10 μg/mL (100 μL/well) on an Nickel Coated plate, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 0.078-5 μg/mL (Routinely tested).