Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Siglec-15 / CD33L3 Protein, His,Avitag™ (recommended for biopanning)
Supplier:  ACROBiosystems SG5H82E9200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human Siglec-15, His,Avitag (recommended for biopanning) (SG5-H82E9) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession # Q6ZMC9-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 30.1 kDa. The protein migrates as 33-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse Human chimeric MAb (5G12, Human IgG1) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Siglec-15, His,Avitag (recommended for biopanning) (Cat. No. SG5-H82E9) with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-195-6840