Learn More
Medchemexpress LLC Siglec-15 Human Bi 100ug
SDP
Catalog No. 5000196985
Encompass_Preferred
Recombinant human Siglec-15 extracellular domain produced in HEK293 cells, biotinylated via an Avi tag and fused to a C-terminal Fc for improved stability and capture. The product is supplied lyophilized with formulation buffer and is intended for use in binding studies, receptor-ligand assays, and immunoassays when reconstituted according to the recommended concentration.
  • Biotinylated Fc-tagged construct for affinity capture and detection.
  • Mammalian expression ensures native glycosylation and folding.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation with protective excipient for stability.
  • Recommended reconstitution at ≥100 μg/mL for optimal activity.
  • Suitable for binding studies, receptor interaction assays, and immunoassays.
Catalog No. 50-001-96985 Supplier Medchemexpress LLC Supplier No. HYP70767100UG
Recombinant human Siglec-15 extracellular domain produced in HEK293 cells, biotinylated via an Avi tag and fused to a C-terminal Fc for improved stability and capture. The product is supplied lyophilized with formulation buffer and is intended for use in binding studies, receptor-ligand assays, and immunoassays when reconstituted according to the recommended concentration.
  • Biotinylated Fc-tagged construct for affinity capture and detection.
  • Mammalian expression ensures native glycosylation and folding.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation with protective excipient for stability.
  • Recommended reconstitution at ≥100 μg/mL for optimal activity.
  • Suitable for binding studies, receptor interaction assays, and immunoassays.