Learn More
Medchemexpress LLC Siglec-15 Human Bi 20ug
SDP
Catalog No. 5000196907
Encompass_Preferred
Recombinant human Siglec-15 extracellular domain expressed in HEK293 cells and provided biotinylated with an Fc-Avi tag. The protein is intended for use in binding assays, receptor-ligand studies, and biochemical characterization requiring a biotinylated mammalian-expressed receptor fragment. Handle and store according to manufacturer recommendations to preserve activity.
  • Biotinylated Fc-Avi tagged extracellular domain for capture and detection.
  • Expressed in HEK293 cells to retain mammalian glycosylation.
  • Supplied as a small-quantity purified protein (20 μg) for small-scale assays.
  • Suitable for binding assays, protein interaction studies, and assay development.
  • Purified and formulated for stability; follow storage instructions on the product page.
Catalog No. 50-001-96907 Supplier Medchemexpress LLC Supplier No. HYP7076720UG
Recombinant human Siglec-15 extracellular domain expressed in HEK293 cells and provided biotinylated with an Fc-Avi tag. The protein is intended for use in binding assays, receptor-ligand studies, and biochemical characterization requiring a biotinylated mammalian-expressed receptor fragment. Handle and store according to manufacturer recommendations to preserve activity.
  • Biotinylated Fc-Avi tagged extracellular domain for capture and detection.
  • Expressed in HEK293 cells to retain mammalian glycosylation.
  • Supplied as a small-quantity purified protein (20 μg) for small-scale assays.
  • Suitable for binding assays, protein interaction studies, and assay development.
  • Purified and formulated for stability; follow storage instructions on the product page.