Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human Siglec-2 Protein, His Tag, Low Endotoxin, 1mg

Catalog No. 501957346
Encompass_Preferred

This Human Siglec-2/CD22 protein is a recombinant protein expressed from human 293 cells. As a member of the immunoglobulin superfamily and the SIGLEC family, it mediates B-cell interactions and acts as an inhibitory receptor. This low-endotoxin protein is suitable for various analytical applications, verified by SEC-MALS and SDS-PAGE.

  • Greater than 95% purity as determined by SDS-PAGE and SEC-MALS.
  • Contains a polyhistidine tag at the C-terminus.
  • Calculated molecular weight of 77.0 kDa, migrating at 90-116 kDa due to glycosylation.
  • Endotoxin level is less than 0.01 EU per μg.
  • Lyophilized from a 0.22 μm filtered PBS solution, pH 7.4, containing trehalose as a protectant.
  • Stable for 12 months at -20°C to -70°C in lyophilized state and 6 months at -70°C after reconstitution under sterile conditions.

Catalog No. 50-195-7346 Supplier ACROBiosystems Supplier No. CD2H52H81MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Human Siglec-2/CD22 protein is a recombinant protein expressed from human 293 cells. As a member of the immunoglobulin superfamily and the SIGLEC family, it mediates B-cell interactions and acts as an inhibitory receptor. This low-endotoxin protein is suitable for various analytical applications, verified by SEC-MALS and SDS-PAGE.

  • Greater than 95% purity as determined by SDS-PAGE and SEC-MALS.
  • Contains a polyhistidine tag at the C-terminus.
  • Calculated molecular weight of 77.0 kDa, migrating at 90-116 kDa due to glycosylation.
  • Endotoxin level is less than 0.01 EU per μg.
  • Lyophilized from a 0.22 μm filtered PBS solution, pH 7.4, containing trehalose as a protectant.
  • Stable for 12 months at -20°C to -70°C in lyophilized state and 6 months at -70°C after reconstitution under sterile conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.