Learn More
ACROBiosystems ACROBiosystems Human Siglec-2 / CD22 Protein, His Tag
Supplier: ACROBiosystems CD2H52H8100UG
Recombinant Protein;100UG;Human Siglec-2, His Tag (CD2-H52H8) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 687 (Accession # P20273-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 77.0 kDa. The protein migrates as 90-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human CD22 Mab at 2 μg/mL (100 μL/well) can bind Human Siglec-2, His Tag (Cat. No. CD2-H52H8) with a linear range of 20-156 ng/mL (QC tested).;293 cells were transfected with anti-CD22-scFv and RFP tag. 2e5 of the cells were first stained with B. Human Siglec-2, His Tag (Cat. No. CD2-H52H8, 3 μg/mL) and C. His Tag Protein Control, followed by FITC Anti-6xHis Tag Antibody.