Learn More
ACROBiosystems ACROBiosystems Human Siglec-3 / CD33 (18-132) Protein, Fc Tag
Supplier:  ACROBiosystems CD3H52551MG
Encompass_Preferred
Recombinant Protein;1MG;Human Siglec-3 (18-132), Fc Tag (CD3-H5255) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - His 132 (Accession # P20138-3).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 38.6 kDa. The protein migrates as 46-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6434