Learn More
ACROBiosystems ACROBiosystems Human Siglec-3 / CD33 Protein, His Tag
Supplier:  ACROBiosystems CD3H5226100UG
Encompass_Preferred
Recombinant Protein;100UG;Human Siglec-3, His Tag (CD3-H5226) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.6 kDa. The protein migrates as 43-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human CD33 MAb, Mouse IgG1 at 2 μg/mL (100 μL/well) can bind Human Siglec-3, His Tag (Cat. No. CD3-H5226) with a linear range of 0.2-1 ng/mL (QC tested).
Catalog No. 50-195-6429