Learn More
ACROBiosystems ACROBiosystems Human Siglec-8 Protein, Fc Tag
Catalog No. 501956758
Encompass_Preferred
Recombinant Protein;1MG;Human Siglec-8, Fc Tag (SI8-H5254) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Ala 363 (Accession # Q9NYZ4-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.3 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6758 Supplier ACROBiosystems Supplier No. SI8H52541MG
Recombinant Protein;1MG;Human Siglec-8, Fc Tag (SI8-H5254) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Ala 363 (Accession # Q9NYZ4-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.3 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.