Learn More
ACROBiosystems ACROBiosystems Human Siglec-8 Protein, His Tag
Catalog No. 501956762
Encompass_Preferred
Recombinant Protein;1MG;Human Siglec-8, His Tag (SI8-H52H8) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Ala 363 (Accession # Q9NYZ4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.7 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6762 Supplier ACROBiosystems Supplier No. SI8H52H81MG
Recombinant Protein;1MG;Human Siglec-8, His Tag (SI8-H52H8) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Ala 363 (Accession # Q9NYZ4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.7 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.