Learn More
ACROBiosystems ACROBiosystems Human Siglec-9 Protein, Fc Tag
Catalog No. 501956710
Encompass_Preferred
Recombinant Protein;1MG;Human Siglec-9, Fc Tag (SI9-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 348 (Accession # Q9Y336-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.5 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-9, Fc Tag (Cat. No. SI9-H5256) at 10 μg/mL (100 μL/well) on Diamond Protein A Protein, His Tag precoated (0.5 μg/well) plate, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 0.156-2.5 μg/mL (QC tested).
Catalog No. 50-195-6710 Supplier ACROBiosystems Supplier No. SI9H52561MG
Recombinant Protein;1MG;Human Siglec-9, Fc Tag (SI9-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 348 (Accession # Q9Y336-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.5 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-9, Fc Tag (Cat. No. SI9-H5256) at 10 μg/mL (100 μL/well) on Diamond Protein A Protein, His Tag precoated (0.5 μg/well) plate, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 0.156-2.5 μg/mL (QC tested).