Learn More
ACROBiosystems ACROBiosystems Cynomolgus Siglec-9 Protein, His Tag
Supplier:  ACROBiosystems SI9C52H91MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus Siglec-9, His Tag (SI9-C52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Gly 350 (Accession # XP_005590140.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 38.0 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus Siglec-9, His Tag (Cat. No. SI9-C52H9) at 10 μg/mL (100 μL/well) on Nickel Coated plate, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 10-156 ng/mL (QC tested).
Catalog No. 50-195-6539