Learn More
ACROBiosystems ACROBiosystems Human Siglec-9 Protein, His Tag
Supplier:  ACROBiosystems SI9H52H41MG
Encompass_Preferred
Recombinant Protein;1MG;Human Siglec-9, His Tag (SI9-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 348 (Accession # Q9Y336-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.9 kDa. The protein migrates as 53-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-9, His Tag (Cat. No. SI9-H52H4) at 10 μg/mL (100 μL/well) on Nickel Coated plate, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 0.078-2.5 μg/mL (QC tested).
Catalog No. 50-195-6714