Learn More
Medchemexpress LLC Siglec-E Mouse HEK 1mg
SDP
Supplier:  Medchemexpress LLC HYP713141MG
Encompass_Preferred
Siglec-E protein is a recombinant mouse extracellular domain fused to a human Fc tag, supplied as a lyophilized powder for use in immunology and cell-signaling research. The construct corresponds to the sequence Q6PJ50 (aa Q20-F355), is produced in HEK293 cells, and shows size heterogeneity from glycosylation. Certificate of analysis reports high purity and provides recommended reconstitution and storage guidance.
  • Recombinant mouse extracellular domain fused to a human Fc tag.
  • Produced in HEK293 expression system for mammalian post-translational modifications.
  • Lyophilized powder formulation for convenient storage and transport.
  • High purity as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~78-100 kDa due to glycosylation.
  • Recommend reconstitution to ≥100 μg/mL in ddH2O.
  • Store lyophilized at -20°C; freeze aliquots at -20°C or -80°C with carrier protein for long-term storage.
Catalog No. 50-001-96891