Learn More
Aviva Systems Biology Corp SIGLEC12 Antibody - N-terminal region (ARP50183_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50183_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIGLEC12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SIGLEC12. Alternate names: FLJ38600, S2V, SIGLECL1, SLG, Siglec-12, Siglec-L1, Siglec-XIIApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rat, Zebrafish
Catalog No. 50-014-60276