Learn More
ACROBiosystems Recombinant Protein;Mouse Siglec-2 / CD22 Protein;HEK293;100UG;SI2,CD22,SIGLEC2,BL-CAM,SIGLEC-2,Siglec2,Siglec-2,SIGLEC2FLJ22814,T-cell surface antigen Leu-14
Catalog No. 501657739
Encompass_Preferred
Recombinant Protein;Mouse Siglec-2 / CD22 Protein;100UG;Mouse Siglec-2 (SI2-M52Ha) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 702 (Accession # P35329-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 78.3 kDa. The protein migrates as 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7739 Supplier ACROBiosystems Supplier No. SI2M52HA100UG
Recombinant Protein;Mouse Siglec-2 / CD22 Protein;100UG;Mouse Siglec-2 (SI2-M52Ha) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 702 (Accession # P35329-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 78.3 kDa. The protein migrates as 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.