Learn More
ACROBiosystems Recombinant Protein;Human Siglec-2 / CD22 Protein, Llama IgG2b Fc Tag, low endotoxin;HEK293;100UG;SI2,CD22,SIGLEC2,BL-CAM,SIGLEC-2,Siglec2,Siglec-2,SIGLEC2FLJ22814,T-cell surface antigen Leu-14
Supplier: ACROBiosystems SI2H525A100UG
Recombinant Protein;Human Siglec-2 / CD22 Protein, Llama IgG2b Fc Tag, low endotoxin;100UG;Human Siglec-2, Llama IgG2b Fc Tag, low endotoxin (SI2-H525a) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 687 (Accession # P20273-1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 103.0 kDa. The protein migrates as 120-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-2, Llama IgG2b Fc Tag, low endotoxin (Cat. No. SI2-H525a) at 2 μg/mL (100 μL/well) can bind Anti-Human CD22 with a linear range of 4-63 ng/mL (QC tested).