Learn More
ACROBiosystems Recombinant Protein;Human Siglec-3 / CD33 Protein, Fc Tag;HEK293;1MG;CD3,CD33,SIGLEC3,SIGLEC-3,gp67
Supplier:  ACROBiosystems CD3H52571MG
Encompass_Preferred
Recombinant Protein;Human Siglec-3 / CD33 Protein, Fc Tag;1MG;Human Siglec-3, Fc Tag (CD3-H5257) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.9 kDa. The protein migrates as 64-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7942