Learn More
ACROBiosystems Recombinant Protein;Human Siglec-5 / CD170 Protein, Fc Tag;HEK293;1MG;SI5,SIGLEC5,OB-BP2,CD170,CD33L2,OBBP2,SIGLEC-5,CD33-L2
Catalog No. 501658545
Encompass_Preferred
Recombinant Protein;Human Siglec-5 / CD170 Protein, Fc Tag;1MG;Human Siglec-5, Fc Tag (SI5-H5250) is expressed from human 293 cells (HEK293). It contains AA Glu 17 - Thr 434 (Accession # AAH29896).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73 kDa. The protein migrates as 90-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8545 Supplier ACROBiosystems Supplier No. SI5H52501MG
Recombinant Protein;Human Siglec-5 / CD170 Protein, Fc Tag;1MG;Human Siglec-5, Fc Tag (SI5-H5250) is expressed from human 293 cells (HEK293). It contains AA Glu 17 - Thr 434 (Accession # AAH29896).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73 kDa. The protein migrates as 90-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.