Learn More
Aviva Systems Biology Corp SIGLEC6 Antibody - N-terminal region (ARP41924_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41924_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIGLEC6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SIGLEC6. Alternate names: CD33L, CD33L1, CDw327, OBBP1, SIGLEC-6, CD327, CD33L2, CDW327ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58046