Learn More
ACROBiosystems Recombinant Protein;Human Siglec-6 / CD327 Protein, Fc Tag;HEK293;1MG;SI6,SIGLEC6,CD327,CD33L,CD33L1,OBBP1,OB-BP1,CDw327,SIGLEC-6,OBBP-1
Supplier:  ACROBiosystems SI6H52561MG
Encompass_Preferred
Recombinant Protein;Human Siglec-6 / CD327 Protein, Fc Tag;1MG;Human Siglec-6, Fc Tag (SI6-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 16 - Val 320 (Accession # AAH35359).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.3 kDa. As a result of glycosylation, the protein migrates as 80-90 kDa under reducing (R) condition, and 160-180 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-8313