Learn More
Aviva Systems Biology Corp SIGLEC7 Antibody - middle region (ARP47017_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47017_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIGLEC7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SIGLEC7. Alternate names: AIRM1, CDw328, D-siglec, QA79, SIGLEC-7, p75, p75/AIRM1, CD328ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59451